Assay Details
Amplicon Length: | 80 |
Chromosome Location: | Chr. 6 - 133860039 - 133860039 [-] on 133860039 Build Rnor_6.0 |
Species: | Rat |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-3579 | MIMAT0017867 |
| Rat | rno-miR-380-5p | MIMAT0005308 |
| Rat | rno-miR-380-3p | MIMAT0017302 |
Stem-loop Details
Rat

Genomic Map