Assay Details
Gene Family ID: | MIPF0000063, mir-192 |
Mature miRNA Sequence: | UCUGUCAUUCUGUAGGCCAAU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-215-3p | MIMAT0017169 |
| Cricetulus griseus | cgr-miR-215-3p | MIMAT0023854 |
Stem-loop Details
Mouse
Cricetulus griseus

Genomic Map