Assay Details
Gene Family ID: | MIPF0000002, let-7 |
Mature miRNA Sequence: | CUAUACAAUCUAUUGCCUUCC |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-let-7f-1-3p | MIMAT0017089 |
| Alligator mississippiensis | ami-let-7f-3p | MIMAT0038013 |
| Anolis carolinensis | aca-let-7f-2-3p | MIMAT0021705 |
| Chrysemys picta | cpi-let-7f-3p | MIMAT0037607 |
| Pig | ssc-let-7f-3p | MIMAT0037464 |
| Python bivittatus | pbv-let-7f-2-3p | MIMAT0038807 |
| View More Rows | ||
| Taeniopygia guttata | tgu-let-7f-3p | MIMAT0026988 |
Stem-loop Details
Rat
Alligator mississippiensis
Anolis carolinensis
Chrysemys picta
Pig
Python bivittatus
Taeniopygia guttata
More Information
Related Products
miRBase Alias:
Rat: rno-let-7f-1* (v18)
Aeromonas caviae Ae398: aca-let-7f-2* (v18)
Aeromonas caviae Ae398: aca-let-7f-2* (v18)

Genomic Map