Assay Details
Amplicon Length: | 72 |
Chromosome Location: | Chr. 17 - 16418570 - 16418570 [-] on 16418570 Build Rnor_6.0 |
Species: | Rat |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Rat | rno-let-7f-5p | MIMAT0000778 |
Rat | rno-let-7f-1-3p | MIMAT0017089 |
Rat | rno-miR-3596d | MIMAT0017823 |
Stem-loop Details
Rat
