Assay Details
Gene Family ID: | MIPF0000111, mir-489 |
Mature miRNA Sequence: | UGUCAUAUGUGUGAUGACACUUUCU |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Mouse | mmu-miR-489-5p | MIMAT0022704 |
Stem-loop Details
Mouse

Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | mmu-miR-489-5p |
---|---|
miRBase Accession Number: | MI0003476 |
miRBase Version: | v22.1 |
Mature miRNA Sequence: | UGUCAUAUGUGUGAUGACACUUUCU |
Species: | Mouse |
Product Type: | Ambion® Anti-miR™ miRNA Inhibitor |
Gene Family ID: | MIPF0000111, mir-489 |
Mature miRNA Sequence: | UGUCAUAUGUGUGAUGACACUUUCU |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Mouse | mmu-miR-489-5p | MIMAT0022704 |