Assay Details
Amplicon Length: | 69 |
Chromosome Location: | Chr. 6 - 3722303 - 3722303 [+] on 3722303 Build GRCm38 |
Species: | Mouse |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Mouse | mmu-miR-489-5p | MIMAT0022704 |
Mouse | mmu-miR-489-3p | MIMAT0003112 |
Stem-loop Details
Mouse
