Assay Details
Amplicon Length: | 90 |
Chromosome Location: | Chr. 9 - 70809778 - 70809778 [-] on 70809778 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-204-5p | MIMAT0000265 |
| Human | hsa-miR-204-3p | MIMAT0022693 |
Stem-loop Details
Human

Genomic Map