Assay Details
Gene Family ID: | MIPF0000042, mir-204 |
Mature miRNA Sequence: | GCUGGGAAGGCAAAGGGACGU |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number | |
|---|---|---|---|
| Human | hsa-miR-204-3p | MIMAT0022693 | |
| Mouse | mmu-miR-204-3p | MIMAT0017002 | |
| Alligator mississippiensis | ami-miR-204-3p | MIMAT0038226 | |
| Anolis carolinensis | aca-miR-204a-3p | MIMAT0021854 | |
| Chrysemys picta | cpi-miR-204-3p | MIMAT0037828 | |
| Columba livia | cli-miR-204-3p | MIMAT0038581 | |
| View More Rows | |||
| Dasypus novemcinctus | dno-miR-204-3p | MIMAT0047745 | |
| Guinea pig | cpo-miR-204-3p | MIMAT0047100 | |
| Ophiophagus hannah | oha-miR-204-1-3p | MIMAT0036813 | |
| Pteropus alecto | pal-miR-204-3p | MIMAT0039966 | |
| Rabbit | ocu-miR-204-3p | MIMAT0048324 | |
| Sarcophilus harrisii | sha-miR-204 | MIMAT0022790 | |
Stem-loop Details
Human
Mouse
Alligator mississippiensis
Anolis carolinensis
Chrysemys picta
Columba livia
Dasypus novemcinctus
Guinea pig
Ophiophagus hannah
Pteropus alecto
Rabbit
Sarcophilus harrisii
More Information
Related Products
miRBase Alias:
Mouse: mmu-miR-204* (v17)
Aeromonas caviae Ae398: aca-miR-204a* (v18)
Aeromonas caviae Ae398: aca-miR-204a* (v18)

Genomic Map