Assay Details
Amplicon Length: | 79 |
Chromosome Location: | Chr. 14 - 101026318 - 101026318 [-] on 101026318 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-758-5p | MIMAT0022929 |
| Human | hsa-miR-758-3p | MIMAT0003879 |
Stem-loop Details
Human

Genomic Map