Assay Details
Gene Family ID: | MIPF0000126, mir-379 |
Mature miRNA Sequence: | UUUGUGACCUGGUCCACUAACC |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-758-3p | MIMAT0003879 |
| Rat | rno-miR-758-3p | MIMAT0005335 |
| Bovine | bta-miR-758 | MIMAT0009370 |
| Dasypus novemcinctus | dno-miR-758-3p | MIMAT0048011 |
| Dog | cfa-miR-758 | MIMAT0009925 |
| Goat | chi-miR-758 | MIMAT0036292 |
| View More Rows | ||
| Gorilla gorilla | ggo-miR-758 | MIMAT0049118 |
| Guinea pig | cpo-miR-758-3p | MIMAT0047382 |
| Horse | eca-miR-758 | MIMAT0013173 |
| Pan troglodytes | ptr-miR-758 | MIMAT0008318 |
| Pongo pygmaeus | ppy-miR-758 | MIMAT0016103 |
| Pteropus alecto | pal-miR-758-3p | MIMAT0040252 |
Stem-loop Details
Human
Rat
Bovine
Dasypus novemcinctus
Dog
Goat
Gorilla gorilla
Guinea pig
Horse
Pan troglodytes
Pongo pygmaeus
Pteropus alecto

Genomic Map