Assay Details
Amplicon Length: | 100 |
Chromosome Location: | Chr. 12 - 54337620 - 54337620 [-] on 54337620 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-148b-3p | MIMAT0000759 |
| Human | hsa-miR-148b-5p | MIMAT0004699 |
Stem-loop Details
Human

Genomic Map