Assay Details
Gene Family ID: | MIPF0000056, mir-148 |
Mature miRNA Sequence: | UCAGUGCAUCACAGAACUUUGU |
Mature miRNA Details:
Species | miRBase ID | miRBase Accession Number | |
---|---|---|---|
Human | hsa-miR-148b-3p | MIMAT0000759 | |
Mouse | mmu-miR-148b-3p | MIMAT0000580 | |
Rat | rno-miR-148b-3p | MIMAT0000579 | |
Anolis carolinensis | aca-miR-148b-3p | MIMAT0021783 | |
Bovine | bta-miR-148b | MIMAT0003814 | |
Callithrix jacchus | cja-miR-148b | MIMAT0039462 | |
View More Rows | |||
Chrysemys picta | cpi-miR-148b-3p | MIMAT0037784 | |
Cricetulus griseus | cgr-miR-148b-3p | MIMAT0023779 | |
Dasypus novemcinctus | dno-miR-148b-3p | MIMAT0047672 | |
Dog | cfa-miR-148b | MIMAT0006663 | |
Goat | chi-miR-148b-3p | MIMAT0035979 | |
Guinea pig | cpo-miR-148b-3p | MIMAT0047025 | |
Horse | eca-miR-148b-3p | MIMAT0012972 | |
Ophiophagus hannah | oha-miR-148b | MIMAT0036735 | |
Pan troglodytes | ptr-miR-148b | MIMAT0008042 | |
Pig | ssc-miR-148b-3p | MIMAT0013901 | |
Pongo pygmaeus | ppy-miR-148b | MIMAT0015772 | |
Pteropus alecto | pal-miR-148b-3p | MIMAT0039869 | |
Python bivittatus | pbv-miR-148b-3p | MIMAT0038967 | |
Rabbit | ocu-miR-148b-3p | MIMAT0048262 | |
Rhesus monkey | mml-miR-148b-3p | MIMAT0006209 | |
Tupaia chinensis | tch-miR-148b-3p | MIMAT0036534 | |
Xenopus tropicalis | xtr-miR-148b | MIMAT0003605 |
Stem-loop Details
Human
Mouse
Rat
Anolis carolinensis
Bovine
Callithrix jacchus
Chrysemys picta
Cricetulus griseus
Dasypus novemcinctus
Dog
Goat
Guinea pig
Horse
Ophiophagus hannah
Pan troglodytes
Pig
Pongo pygmaeus
Pteropus alecto
Python bivittatus
Rabbit
Rhesus monkey
Tupaia chinensis
Xenopus tropicalis

More Information
Associated Megaplex™ RT Primer Pools:
Megaplex™ RT Primers, Rodent Pool AMegaplex™ RT Primers, Human Pool A
Related Products
miRBase Alias:
Human: hsa-miR-148b (v17)
Mouse: mmu-miR-148b (v17)
Rat: rno-miR-148b (v9.2)
Aeromonas caviae Ae398: aca-miR-148b (v18)
Macaca mulatta: mml-miR-148b (v19)
Staphylococcus schleiferi: ssc-miR-148b (v18)
Mouse: mmu-miR-148b (v17)
Rat: rno-miR-148b (v9.2)
Aeromonas caviae Ae398: aca-miR-148b (v18)
Macaca mulatta: mml-miR-148b (v19)
Staphylococcus schleiferi: ssc-miR-148b (v18)
