Assay Details
Amplicon Length: | 59 |
Chromosome Location: | Chr. X - 74286907 - 74286907 [+] on 74286907 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-545-5p | MIMAT0004785 |
| Human | hsa-miR-545-3p | MIMAT0003165 |
Stem-loop Details
Human

Genomic Map