Assay Details
Gene Family ID: | MIPF0000098, mir-95 |
Mature miRNA Sequence: | UCAGCAAACAUUUAUUGUGUGC |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-545-3p | MIMAT0003165 |
| Pan troglodytes | ptr-miR-545 | MIMAT0008217 |
| Pongo pygmaeus | ppy-miR-545 | MIMAT0015995 |
| Rhesus monkey | mml-miR-545 | MIMAT0006409 |
Stem-loop Details
Human
Pan troglodytes
Pongo pygmaeus
Rhesus monkey

Genomic Map