Assay Details
Amplicon Length: | 51 |
Chromosome Location: | Chr. 14 - 101560686 - 101560686 [+] on 101560686 Build GRCh38 |
Species: | Human |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-1247-3p | MIMAT0022721 |
Human | hsa-miR-1247-5p | MIMAT0005899 |
Stem-loop Details
Human
