Assay Details
Gene Family ID: | MIPF0000669, mir-1247 |
Mature miRNA Sequence: | ACCCGUCCCGUUCGUCCCCGGA |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-1247-5p | MIMAT0005899 |
| Mouse | mmu-miR-1247-5p | MIMAT0014800 |
| Rat | rno-miR-1247-5p | MIMAT0035721 |
| Guinea pig | cpo-miR-1247-5p | MIMAT0047405 |
| Pan troglodytes | ptr-miR-1247 | MIMAT0007973 |
| Pongo pygmaeus | ppy-miR-1247 | MIMAT0016172 |
| View More Rows | ||
| Rhesus monkey | mml-miR-1247-5p | MIMAT0028358 |
| Tupaia chinensis | tch-miR-1247-5p | MIMAT0036594 |
Stem-loop Details
Human
Mouse
Rat
Guinea pig
Pan troglodytes
Pongo pygmaeus
Rhesus monkey
Tupaia chinensis

Genomic Map