Assay Details
Amplicon Length: | 88 |
Chromosome Location: | Chr. 1 - 68183216 - 68183216 [-] on 68183216 Build GRCh38 |
Species: | Human |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-1262 | MIMAT0005914 |
Stem-loop Details
Human
