Assay Details
Gene Family ID: | MIPF0000595, mir-1262 |
Mature miRNA Sequence: | AUGGGUGAAUUUGUAGAAGGAU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-1262 | MIMAT0005914 |
| Pan troglodytes | ptr-miR-1262 | MIMAT0007986 |
| Pongo pygmaeus | ppy-miR-1262 | MIMAT0016186 |
Stem-loop Details
Human
Pan troglodytes
Pongo pygmaeus

Genomic Map