Assay Details
Amplicon Length: | 88 |
Chromosome Location: | Chr. 15 - 52276715 - 52276715 [+] on 52276715 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-1266-3p | MIMAT0026742 |
| Human | hsa-miR-1266-5p | MIMAT0005920 |
Stem-loop Details
Human

Genomic Map