Assay Details
Gene Family ID: | MIPF0000615, mir-1266 |
Mature miRNA Sequence: | CCUCAGGGCUGUAGAACAGGGCU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-1266-5p | MIMAT0005920 |
| Pan troglodytes | ptr-miR-1266 | MIMAT0007990 |
Stem-loop Details
Human
Pan troglodytes

Genomic Map