Assay Details
Amplicon Length: | 85 |
Chromosome Location: | Chr. X - 118386195 - 118386195 [+] on 118386195 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-1277-3p | MIMAT0005933 |
| Human | hsa-miR-1277-5p | MIMAT0022724 |
Stem-loop Details
Human

Genomic Map