Assay Details
Gene Family ID: | MIPF0001937, mir-1277 |
Mature miRNA Sequence: | UACGUAGAUAUAUAUGUAUUUU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-1277-3p | MIMAT0005933 |
| Bovine | bta-miR-1277 | MIMAT0024569 |
| Dasypus novemcinctus | dno-miR-1277-3p | MIMAT0048038 |
| Guinea pig | cpo-miR-1277-3p | MIMAT0047414 |
| Pig | ssc-miR-1277 | MIMAT0013935 |
| Pongo pygmaeus | ppy-miR-1277 | MIMAT0016200 |
| View More Rows | ||
| Rabbit | ocu-miR-1277-3p | MIMAT0048592 |
Stem-loop Details
Human
Bovine
Dasypus novemcinctus
Guinea pig
Pig
Pongo pygmaeus
Rabbit

Genomic Map