Assay Details
Amplicon Length: | 110 |
Chromosome Location: | Chr. 21 - 18686334 - 18686334 [-] on 18686334 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-548x-3p | MIMAT0015081 |
| Human | hsa-miR-548x-5p | MIMAT0022733 |
Stem-loop Details
Human

Genomic Map