Assay Details
Gene Family ID: | MIPF0000317, mir-548 |
Mature miRNA Sequence: | UGCAAAAGUAAUUGCAGUUUUUG |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-548aj-5p | MIMAT0022739 |
| Human | hsa-miR-548g-5p | MIMAT0022722 |
| Human | hsa-miR-548x-5p | MIMAT0022733 |
Stem-loop Details
Human

Genomic Map