Assay Details
Amplicon Length: | 105 |
Chromosome Location: | Chr. X - 38023644 - 38023644 [-] on 38023644 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-548aj-3p | MIMAT0018990 |
| Human | hsa-miR-548aj-5p | MIMAT0022739 |
Stem-loop Details
Human

Genomic Map