Assay Details
Gene Family ID: | MIPF0000165, mir-324 |
Mature miRNA Sequence: | CCACUGCCCCAGGUGCUGCUGG |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Rat | rno-miR-324-3p | MIMAT0000554 |
Cricetulus griseus | cgr-miR-324-3p | MIMAT0023910 |
Dasypus novemcinctus | dno-miR-324-3p | MIMAT0047804 |
Eptesicus fuscus | efu-miR-324 | MIMAT0035051 |
Guinea pig | cpo-miR-324-3p | MIMAT0047159 |
Horse | eca-miR-324-3p | MIMAT0013034 |
View More Rows | ||
Rabbit | ocu-miR-324-3p | MIMAT0048371 |
Stem-loop Details
Rat
Cricetulus griseus
Dasypus novemcinctus
Eptesicus fuscus
Guinea pig
Horse
Rabbit
