Assay Details
Amplicon Length: | 72 |
Chromosome Location: | Chr. 10 - 56621383 - 56621383 [-] on 56621383 Build Rnor_6.0 |
Species: | Rat |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Rat | rno-miR-324-3p | MIMAT0000554 |
Rat | rno-miR-324-5p | MIMAT0000553 |
Stem-loop Details
Rat
