Assay Details
| Mature miRNA Sequence: | UAUGGCACUGGUAGAAUUCACUG | 
miRBase Details:
| Species | miRBase ID | miRBase Accession Number | 
|---|---|---|
| Astatotilapia burtoni | abu-miR-183 | MIMAT0042040 | 
| Bovine | bta-miR-183 | MIMAT0009245 | 
| Chicken | gga-miR-183 | MIMAT0001191 | 
| Ciona intestinalis | cin-miR-183-5p | MIMAT0006107 | 
| Columba livia | cli-miR-183-5p | MIMAT0038558 | 
| Cyprinus carpio | ccr-miR-183 | MIMAT0026243 | 
| View More Rows | ||
| Fugu rubripes | fru-miR-183 | MIMAT0003089 | 
| Gorilla gorilla | ggo-miR-183 | MIMAT0002787 | 
| Macaca nemestrina | mne-miR-183 | MIMAT0002791 | 
| Metriaclima zebra | mze-miR-183 | MIMAT0042279 | 
| Monodelphis domestica | mdo-miR-183-5p | MIMAT0004146 | 
| Neolamprologus brichardi | nbr-miR-183 | MIMAT0042467 | 
| Ophiophagus hannah | oha-miR-183-5p | MIMAT0036766 | 
| Oreochromis niloticus | oni-miR-183 | MIMAT0042645 | 
| Pan paniscus | ppa-miR-183 | MIMAT0002793 | 
| Pan troglodytes | ptr-miR-183 | MIMAT0002785 | 
| Petromyzon marinus | pma-miR-183-5p | MIMAT0019491 | 
| Pig | ssc-miR-183 | MIMAT0002145 | 
| Pundamilia nyererei | pny-miR-183 | MIMAT0042855 | 
| Python bivittatus | pbv-miR-183-5p | MIMAT0038982 | 
| Rhesus monkey | mml-miR-183-5p | MIMAT0002783 | 
| Saguinus labiatus | sla-miR-183 | MIMAT0002789 | 
| Tetraodon nigroviridis | tni-miR-183 | MIMAT0003090 | 
| Xenopus laevis | xla-miR-183-5p | MIMAT0046493 | 
| Xenopus tropicalis | xtr-miR-183 | MIMAT0003612 | 
| Zebrafish | dre-miR-183-5p | MIMAT0001273 | 
Stem-loop Details
Astatotilapia burtoni
| Stem-loop ID | abu-mir-183-1 | 
| Stem-loop Accession # | MI0033715 | 
| Stem-loop Sequence | UAUGGCACUGGUAGAAUUCACUGUCACAGCACACUAUCAGUGAAUUACCAUAGGGCCAUAAA | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0042040 miRBase ID: abu-miR-183 Mature miRNA Sequence: UAUGGCACUGGUAGAAUUCACUG Chromosome Location: NA | 
| Stem-loop ID | abu-mir-183-2 | 
| Stem-loop Accession # | MI0033714 | 
| Stem-loop Sequence | UAUGGCACUGGUAGAAUUCACUGUGAGAGCUCACUAUCAGUGAAUUACCAUAGGGCCAUAAA | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0042040 miRBase ID: abu-miR-183 Mature miRNA Sequence: UAUGGCACUGGUAGAAUUCACUG Chromosome Location: NA | 
| Stem-loop ID | bta-mir-183 | 
| Stem-loop Accession # | MI0009756 | 
| Stem-loop Sequence | CUGUGUAUGGCACUGGUAGAAUUCACUGUGAACAGUCUCGGUCAGUGAAUUACCGAAGGGCCAUAAACAG | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0009245 miRBase ID: bta-miR-183 Mature miRNA Sequence: UAUGGCACUGGUAGAAUUCACUG Chromosome Location: NA | 
| Stem-loop ID | gga-mir-183 | 
| Stem-loop Accession # | MI0001278 | 
| Stem-loop Sequence | ACUCCUGUUCUGUGUAUGGCACUGGUAGAAUUCACUGUGCAACCCCGCGGUCAGUGAAUUACCAUAGGGCCAUAAACAGAGCAGAGAAAGACCCGCGA | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0001191 miRBase ID: gga-miR-183 Mature miRNA Sequence: UAUGGCACUGGUAGAAUUCACUG Chromosome Location: NA | 
| Stem-loop ID | cin-mir-183 | 
| Stem-loop Accession # | MI0007172 | 
| Stem-loop Sequence | CUGGUGUUUUCGGUGUAUGGCACUGGUAGAAUUCACUGCCAAGCCAGUGGUUCAACAAGCGCCGUAAGCUGGAAACACUA | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0006107 miRBase ID: cin-miR-183-5p Mature miRNA Sequence: UAUGGCACUGGUAGAAUUCACUG Chromosome Location: NA | 
| Stem-loop ID | cli-mir-183 | 
| Stem-loop Accession # | MI0029994 | 
| Stem-loop Sequence | UACUCCUGUUCUGUGUAUGGCACUGGUAGAAUUCACUGUGCAACCCCGCGGUCAGUGAAUUACCAUAGGGCCAUAAACAGAGCAGAGAAAG | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0038558 miRBase ID: cli-miR-183-5p Mature miRNA Sequence: UAUGGCACUGGUAGAAUUCACUG Chromosome Location: NA | 
| Stem-loop ID | ccr-mir-183 | 
| Stem-loop Accession # | MI0023347 | 
| Stem-loop Sequence | UGUUCUGUGUAUGGCACUGGUAGAAUUCACUGUGAAAGCACACUAUCAGUGAAUUACCAAAGGGCCAUAAACAGAGCAGA | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0026243 miRBase ID: ccr-miR-183 Mature miRNA Sequence: UAUGGCACUGGUAGAAUUCACUG Chromosome Location: NA | 
| Stem-loop ID | fru-mir-183 | 
| Stem-loop Accession # | MI0003453 | 
| Stem-loop Sequence | CUGUGUAUGGCACUGGUAGAAUUCACUGUGAGAGCUCACUAUCAGUGAAUUACCAUAGGGCCAUAAACAG | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0003089 miRBase ID: fru-miR-183 Mature miRNA Sequence: UAUGGCACUGGUAGAAUUCACUG Chromosome Location: NA | 
| Stem-loop ID | ggo-mir-183 | 
| Stem-loop Accession # | MI0003088 | 
| Stem-loop Sequence | CCGCAGAGUGUGACUCCUGUUCUGUGUAUGGCACUGGUAGAAUUCACUGUGAACAGUCUCAGUCAGUGAAUUACCGAAGGGCCAUAAACAGAGCAGAGACAGAUCCACGA | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0002787 miRBase ID: ggo-miR-183 Mature miRNA Sequence: UAUGGCACUGGUAGAAUUCACUG Chromosome Location: NA | 
| Stem-loop ID | mne-mir-183 | 
| Stem-loop Accession # | MI0003092 | 
| Stem-loop Sequence | CCGCAGAACGUGACUCCUGUUCUGUGUAUGGCACUGGUAGAAUUCACUGUGAACAGUCUCGGUCAGUGAAUUACCGAAGGGCCAUAAACAGAGCAGAGACAGAUCCACGA | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0002791 miRBase ID: mne-miR-183 Mature miRNA Sequence: UAUGGCACUGGUAGAAUUCACUG Chromosome Location: NA | 
| Stem-loop ID | mze-mir-183-1 | 
| Stem-loop Accession # | MI0034015 | 
| Stem-loop Sequence | UAUGGCACUGGUAGAAUUCACUGUCACAGCACACUAUCAGUGAAUUACCAUAGGGCCAUAAA | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0042279 miRBase ID: mze-miR-183 Mature miRNA Sequence: UAUGGCACUGGUAGAAUUCACUG Chromosome Location: NA | 
| Stem-loop ID | mze-mir-183-2 | 
| Stem-loop Accession # | MI0034014 | 
| Stem-loop Sequence | UAUGGCACUGGUAGAAUUCACUGUGAGAGCUCACUAUCAGUGAAUUACCAUAGGGCCAUAAA | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0042279 miRBase ID: mze-miR-183 Mature miRNA Sequence: UAUGGCACUGGUAGAAUUCACUG Chromosome Location: NA | 
| Stem-loop ID | mdo-mir-183 | 
| Stem-loop Accession # | MI0005335 | 
| Stem-loop Sequence | ACUCCUGUUCUGUGUAUGGCACUGGUAGAAUUCACUGUGAAAACACACUAUCAGUGAAUUACCAAAGGGCCAUAAACAGAGCAGAGAA | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0004146 miRBase ID: mdo-miR-183-5p Mature miRNA Sequence: UAUGGCACUGGUAGAAUUCACUG Chromosome Location: NA | 
| Stem-loop ID | nbr-mir-183-1 | 
| Stem-loop Accession # | MI0034276 | 
| Stem-loop Sequence | UAUGGCACUGGUAGAAUUCACUGUCACAGCACACUAUCAGUGAAUUACCAUAGGGCCAUAAA | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0042467 miRBase ID: nbr-miR-183 Mature miRNA Sequence: UAUGGCACUGGUAGAAUUCACUG Chromosome Location: NA | 
| Stem-loop ID | nbr-mir-183-2 | 
| Stem-loop Accession # | MI0034275 | 
| Stem-loop Sequence | UAUGGCACUGGUAGAAUUCACUGUGAGAGCUCACUAUCAGUGAAUUACCAUAGGGCCAUAAA | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0042467 miRBase ID: nbr-miR-183 Mature miRNA Sequence: UAUGGCACUGGUAGAAUUCACUG Chromosome Location: NA | 
| Stem-loop ID | oha-mir-183 | 
| Stem-loop Accession # | MI0031380 | 
| Stem-loop Sequence | CUGUUCUGUGUAUGGCACUGGUAGAAUUCACUGUGAAAACACUCGAUCAGUGAAUUACAAAGGGCCAUAAACGGAG | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0036766 miRBase ID: oha-miR-183-5p Mature miRNA Sequence: UAUGGCACUGGUAGAAUUCACUG Chromosome Location: NA | 
| Stem-loop ID | oni-mir-183-1 | 
| Stem-loop Accession # | MI0034523 | 
| Stem-loop Sequence | UAUGGCACUGGUAGAAUUCACUGUCACAGCACACUAUCAGUGAAUUACCAUAGGGCCAUAAA | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0042645 miRBase ID: oni-miR-183 Mature miRNA Sequence: UAUGGCACUGGUAGAAUUCACUG Chromosome Location: NA | 
| Stem-loop ID | oni-mir-183-2 | 
| Stem-loop Accession # | MI0034522 | 
| Stem-loop Sequence | UAUGGCACUGGUAGAAUUCACUGUGAGAGCUCACUAUCAGUGAAUUACCAUAGGGCCAUAAA | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0042645 miRBase ID: oni-miR-183 Mature miRNA Sequence: UAUGGCACUGGUAGAAUUCACUG Chromosome Location: NA | 
| Stem-loop ID | oni-mir-183-3 | 
| Stem-loop Accession # | MI0035058 | 
| Stem-loop Sequence | UAUGGCACUGGUAGAAUUCACUGUCACAGCACACUAUCAGUGAAUUACCAUAGGGCCAUA | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0042645 miRBase ID: oni-miR-183 Mature miRNA Sequence: UAUGGCACUGGUAGAAUUCACUG Chromosome Location: NA | 
| Stem-loop ID | ppa-mir-183 | 
| Stem-loop Accession # | MI0003094 | 
| Stem-loop Sequence | CCGCAGAGUGUGACUCCUGUUCUGUGUAUGGCACUGGUAGAAUUCACUGUGAACAGUCUCAGUCAGUGAAUUACCGAAGGGCCAUAAACAGAGCAGAGACAGAUCCACGA | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0002793 miRBase ID: ppa-miR-183 Mature miRNA Sequence: UAUGGCACUGGUAGAAUUCACUG Chromosome Location: NA | 
| Stem-loop ID | ptr-mir-183 | 
| Stem-loop Accession # | MI0003086 | 
| Stem-loop Sequence | CCGCAGAGUGUGACUCCUGUUCUGUGUAUGGCACUGGUAGAAUUCACUGUGAACAGUCUCAGUCAGUGAAUUACCGAAGGGCCAUAAACAGAGCAGAGACAGAUCCACGA | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0002785 miRBase ID: ptr-miR-183 Mature miRNA Sequence: UAUGGCACUGGUAGAAUUCACUG Chromosome Location: NA | 
| Stem-loop ID | pma-mir-183 | 
| Stem-loop Accession # | MI0017092 | 
| Stem-loop Sequence | CUGCCUGCCCUGUCUAUGGCACUGGUAGAAUUCACUGUUCCCGCUGACCUCAGUGAAUUACUAGAGGCCAUAAACAGAGCAGGGAA | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0019491 miRBase ID: pma-miR-183-5p Mature miRNA Sequence: UAUGGCACUGGUAGAAUUCACUG Chromosome Location: NA | 
| Stem-loop ID | ssc-mir-183 | 
| Stem-loop Accession # | MI0002439 | 
| Stem-loop Sequence | CUGUGUAUGGCACUGGUAGAAUUCACUGUGAACAGUCUCGGUCAGUGAAUUACCGAAGGGCCAUAAACAG | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0002145 miRBase ID: ssc-miR-183 Mature miRNA Sequence: UAUGGCACUGGUAGAAUUCACUG Chromosome Location: NA | 
| Stem-loop ID | pny-mir-183-1 | 
| Stem-loop Accession # | MI0034795 | 
| Stem-loop Sequence | UAUGGCACUGGUAGAAUUCACUGUCACAGCACACUAUCAGUGAAUUACCAUAGGGCCAUAAA | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0042855 miRBase ID: pny-miR-183 Mature miRNA Sequence: UAUGGCACUGGUAGAAUUCACUG Chromosome Location: NA | 
| Stem-loop ID | pny-mir-183-2 | 
| Stem-loop Accession # | MI0034794 | 
| Stem-loop Sequence | UAUGGCACUGGUAGAAUUCACUGUGAGAGCUCACUAUCAGUGAAUUACCAUAGGGCCAUAAA | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0042855 miRBase ID: pny-miR-183 Mature miRNA Sequence: UAUGGCACUGGUAGAAUUCACUG Chromosome Location: NA | 
| Stem-loop ID | pbv-mir-183 | 
| Stem-loop Accession # | MI0030260 | 
| Stem-loop Sequence | UCCUGUUCUGUGUAUGGCACUGGUAGAAUUCACUGUGAAAACACUCGAUCAGUGAAUUACCAACGGGCCAUAAACGGAGCAGAGAAAGAACC | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0038982 miRBase ID: pbv-miR-183-5p Mature miRNA Sequence: UAUGGCACUGGUAGAAUUCACUG Chromosome Location: NA | 
| Stem-loop ID | mml-mir-183 | 
| Stem-loop Accession # | MI0003084 | 
| Stem-loop Sequence | CCGCAGAACGUGACUCCUGUUCUGUGUAUGGCACUGGUAGAAUUCACUGUGAACAGUCUCGGUCAGUGAAUUACCGAAGGGCCAUAAACAGAGCAGAGACAGAUCCACGA | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0002783 miRBase ID: mml-miR-183-5p Mature miRNA Sequence: UAUGGCACUGGUAGAAUUCACUG Chromosome Location: NA | 
| Stem-loop ID | sla-mir-183 | 
| Stem-loop Accession # | MI0003090 | 
| Stem-loop Sequence | CCGCAGAACGUGACUCCUGUUCUGUGUAUGGCACUGGUAGAAUUCACUGUGAACAGUCUCGGUCAGUGAAUUACCGAAGGGCCAUAAACAGAGCAGAGACAGAUCCUCGA | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0002789 miRBase ID: sla-miR-183 Mature miRNA Sequence: UAUGGCACUGGUAGAAUUCACUG Chromosome Location: NA | 
| Stem-loop ID | tni-mir-183 | 
| Stem-loop Accession # | MI0003454 | 
| Stem-loop Sequence | CUGUGUAUGGCACUGGUAGAAUUCACUGUGAGAGCUCACUAUCAGUGAAUUACCAUAGGGCCAUAAACAG | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0003090 miRBase ID: tni-miR-183 Mature miRNA Sequence: UAUGGCACUGGUAGAAUUCACUG Chromosome Location: NA | 
| Stem-loop ID | xla-mir-183 | 
| Stem-loop Accession # | MI0038274 | 
| Stem-loop Sequence | UAUGGCACUGGUAGAAUUCACUGUGAAAACACAAAAUCAGUGAAUUACCAUAGGGCCAUAAA | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0046493 miRBase ID: xla-miR-183-5p Mature miRNA Sequence: UAUGGCACUGGUAGAAUUCACUG Chromosome Location: NA | 
| Stem-loop ID | xtr-mir-183 | 
| Stem-loop Accession # | MI0004852 | 
| Stem-loop Sequence | CAUCCCUCCUGUUCUGUGUAUGGCACUGGUAGAAUUCACUGUGAAAACACAAAAUCAGUGAAUUACCAUAGGGCCAUAAACAGAGCAGAGAAAGAAC | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0003612 miRBase ID: xtr-miR-183 Mature miRNA Sequence: UAUGGCACUGGUAGAAUUCACUG Chromosome Location: NA | 
| Stem-loop ID | dre-mir-183 | 
| Stem-loop Accession # | MI0001369 | 
| Stem-loop Sequence | GACUCCUGUUCUGUGUAUGGCACUGGUAGAAUUCACUGUGAAAGCACACUAUCAGUGAAUUACCAAAGGGCCAUAAACAGAGCAGAGAAAGAACCACG | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0001273 miRBase ID: dre-miR-183-5p Mature miRNA Sequence: UAUGGCACUGGUAGAAUUCACUG Chromosome Location: NA | 
 Back To Top
 Back To TopMore Information
Related Products
 Ambion® Anti-miR™ miRNA Inhibitor : AM10415
						Ambion® Anti-miR™ miRNA Inhibitor : AM10415 Ambion® Pre-miR™ miRNA Precursor : PM10415
						Ambion® Pre-miR™ miRNA Precursor : PM10415 TaqMan™ MicroRNA Assay : 000484
						TaqMan™ MicroRNA Assay : 000484 mirVana® miRNA inhibitor : MH10415
						mirVana® miRNA inhibitor : MH10415miRBase Alias:
						
							Chryseobacterium indologenes: cin-miR-183 (v18)
Danio rerio: dre-miR-183 (v20)
Macaca mulatta: mml-miR-183 (v19)
Monodelphis domestica: mdo-miR-183 (v19)
Phocea massiliensis: pma-miR-183 (v18)
						
					
				Danio rerio: dre-miR-183 (v20)
Macaca mulatta: mml-miR-183 (v19)
Monodelphis domestica: mdo-miR-183 (v19)
Phocea massiliensis: pma-miR-183 (v18)
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top