Assay Details
Mature miRNA Sequence: | UAUGGCACUGGUAGAAUUCACUG |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Astatotilapia burtoni | abu-miR-183 | MIMAT0042040 |
| Bovine | bta-miR-183 | MIMAT0009245 |
| Chicken | gga-miR-183 | MIMAT0001191 |
| Ciona intestinalis | cin-miR-183-5p | MIMAT0006107 |
| Columba livia | cli-miR-183-5p | MIMAT0038558 |
| Cyprinus carpio | ccr-miR-183 | MIMAT0026243 |
| View More Rows | ||
| Fugu rubripes | fru-miR-183 | MIMAT0003089 |
| Gorilla gorilla | ggo-miR-183 | MIMAT0002787 |
| Macaca nemestrina | mne-miR-183 | MIMAT0002791 |
| Metriaclima zebra | mze-miR-183 | MIMAT0042279 |
| Monodelphis domestica | mdo-miR-183-5p | MIMAT0004146 |
| Neolamprologus brichardi | nbr-miR-183 | MIMAT0042467 |
| Ophiophagus hannah | oha-miR-183-5p | MIMAT0036766 |
| Oreochromis niloticus | oni-miR-183 | MIMAT0042645 |
| Pan paniscus | ppa-miR-183 | MIMAT0002793 |
| Pan troglodytes | ptr-miR-183 | MIMAT0002785 |
| Petromyzon marinus | pma-miR-183-5p | MIMAT0019491 |
| Pig | ssc-miR-183 | MIMAT0002145 |
| Pundamilia nyererei | pny-miR-183 | MIMAT0042855 |
| Python bivittatus | pbv-miR-183-5p | MIMAT0038982 |
| Rhesus monkey | mml-miR-183-5p | MIMAT0002783 |
| Saguinus labiatus | sla-miR-183 | MIMAT0002789 |
| Tetraodon nigroviridis | tni-miR-183 | MIMAT0003090 |
| Xenopus laevis | xla-miR-183-5p | MIMAT0046493 |
| Xenopus tropicalis | xtr-miR-183 | MIMAT0003612 |
| Zebrafish | dre-miR-183-5p | MIMAT0001273 |
Stem-loop Details
Astatotilapia burtoni
| Stem-loop ID | abu-mir-183-1 |
| Stem-loop Accession # | MI0033715 |
| Stem-loop Sequence | UAUGGCACUGGUAGAAUUCACUGUCACAGCACACUAUCAGUGAAUUACCAUAGGGCCAUAAA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0042040
miRBase ID: abu-miR-183 Mature miRNA Sequence: UAUGGCACUGGUAGAAUUCACUG Chromosome Location: NA |
| Stem-loop ID | abu-mir-183-2 |
| Stem-loop Accession # | MI0033714 |
| Stem-loop Sequence | UAUGGCACUGGUAGAAUUCACUGUGAGAGCUCACUAUCAGUGAAUUACCAUAGGGCCAUAAA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0042040
miRBase ID: abu-miR-183 Mature miRNA Sequence: UAUGGCACUGGUAGAAUUCACUG Chromosome Location: NA |
| Stem-loop ID | bta-mir-183 |
| Stem-loop Accession # | MI0009756 |
| Stem-loop Sequence | CUGUGUAUGGCACUGGUAGAAUUCACUGUGAACAGUCUCGGUCAGUGAAUUACCGAAGGGCCAUAAACAG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0009245
miRBase ID: bta-miR-183 Mature miRNA Sequence: UAUGGCACUGGUAGAAUUCACUG Chromosome Location: NA |
| Stem-loop ID | gga-mir-183 |
| Stem-loop Accession # | MI0001278 |
| Stem-loop Sequence | ACUCCUGUUCUGUGUAUGGCACUGGUAGAAUUCACUGUGCAACCCCGCGGUCAGUGAAUUACCAUAGGGCCAUAAACAGAGCAGAGAAAGACCCGCGA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0001191
miRBase ID: gga-miR-183 Mature miRNA Sequence: UAUGGCACUGGUAGAAUUCACUG Chromosome Location: NA |
| Stem-loop ID | cin-mir-183 |
| Stem-loop Accession # | MI0007172 |
| Stem-loop Sequence | CUGGUGUUUUCGGUGUAUGGCACUGGUAGAAUUCACUGCCAAGCCAGUGGUUCAACAAGCGCCGUAAGCUGGAAACACUA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0006107
miRBase ID: cin-miR-183-5p Mature miRNA Sequence: UAUGGCACUGGUAGAAUUCACUG Chromosome Location: NA |
| Stem-loop ID | cli-mir-183 |
| Stem-loop Accession # | MI0029994 |
| Stem-loop Sequence | UACUCCUGUUCUGUGUAUGGCACUGGUAGAAUUCACUGUGCAACCCCGCGGUCAGUGAAUUACCAUAGGGCCAUAAACAGAGCAGAGAAAG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0038558
miRBase ID: cli-miR-183-5p Mature miRNA Sequence: UAUGGCACUGGUAGAAUUCACUG Chromosome Location: NA |
| Stem-loop ID | ccr-mir-183 |
| Stem-loop Accession # | MI0023347 |
| Stem-loop Sequence | UGUUCUGUGUAUGGCACUGGUAGAAUUCACUGUGAAAGCACACUAUCAGUGAAUUACCAAAGGGCCAUAAACAGAGCAGA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0026243
miRBase ID: ccr-miR-183 Mature miRNA Sequence: UAUGGCACUGGUAGAAUUCACUG Chromosome Location: NA |
| Stem-loop ID | fru-mir-183 |
| Stem-loop Accession # | MI0003453 |
| Stem-loop Sequence | CUGUGUAUGGCACUGGUAGAAUUCACUGUGAGAGCUCACUAUCAGUGAAUUACCAUAGGGCCAUAAACAG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0003089
miRBase ID: fru-miR-183 Mature miRNA Sequence: UAUGGCACUGGUAGAAUUCACUG Chromosome Location: NA |
| Stem-loop ID | ggo-mir-183 |
| Stem-loop Accession # | MI0003088 |
| Stem-loop Sequence | CCGCAGAGUGUGACUCCUGUUCUGUGUAUGGCACUGGUAGAAUUCACUGUGAACAGUCUCAGUCAGUGAAUUACCGAAGGGCCAUAAACAGAGCAGAGACAGAUCCACGA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0002787
miRBase ID: ggo-miR-183 Mature miRNA Sequence: UAUGGCACUGGUAGAAUUCACUG Chromosome Location: NA |
| Stem-loop ID | mne-mir-183 |
| Stem-loop Accession # | MI0003092 |
| Stem-loop Sequence | CCGCAGAACGUGACUCCUGUUCUGUGUAUGGCACUGGUAGAAUUCACUGUGAACAGUCUCGGUCAGUGAAUUACCGAAGGGCCAUAAACAGAGCAGAGACAGAUCCACGA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0002791
miRBase ID: mne-miR-183 Mature miRNA Sequence: UAUGGCACUGGUAGAAUUCACUG Chromosome Location: NA |
| Stem-loop ID | mze-mir-183-1 |
| Stem-loop Accession # | MI0034015 |
| Stem-loop Sequence | UAUGGCACUGGUAGAAUUCACUGUCACAGCACACUAUCAGUGAAUUACCAUAGGGCCAUAAA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0042279
miRBase ID: mze-miR-183 Mature miRNA Sequence: UAUGGCACUGGUAGAAUUCACUG Chromosome Location: NA |
| Stem-loop ID | mze-mir-183-2 |
| Stem-loop Accession # | MI0034014 |
| Stem-loop Sequence | UAUGGCACUGGUAGAAUUCACUGUGAGAGCUCACUAUCAGUGAAUUACCAUAGGGCCAUAAA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0042279
miRBase ID: mze-miR-183 Mature miRNA Sequence: UAUGGCACUGGUAGAAUUCACUG Chromosome Location: NA |
| Stem-loop ID | mdo-mir-183 |
| Stem-loop Accession # | MI0005335 |
| Stem-loop Sequence | ACUCCUGUUCUGUGUAUGGCACUGGUAGAAUUCACUGUGAAAACACACUAUCAGUGAAUUACCAAAGGGCCAUAAACAGAGCAGAGAA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0004146
miRBase ID: mdo-miR-183-5p Mature miRNA Sequence: UAUGGCACUGGUAGAAUUCACUG Chromosome Location: NA |
| Stem-loop ID | nbr-mir-183-1 |
| Stem-loop Accession # | MI0034276 |
| Stem-loop Sequence | UAUGGCACUGGUAGAAUUCACUGUCACAGCACACUAUCAGUGAAUUACCAUAGGGCCAUAAA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0042467
miRBase ID: nbr-miR-183 Mature miRNA Sequence: UAUGGCACUGGUAGAAUUCACUG Chromosome Location: NA |
| Stem-loop ID | nbr-mir-183-2 |
| Stem-loop Accession # | MI0034275 |
| Stem-loop Sequence | UAUGGCACUGGUAGAAUUCACUGUGAGAGCUCACUAUCAGUGAAUUACCAUAGGGCCAUAAA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0042467
miRBase ID: nbr-miR-183 Mature miRNA Sequence: UAUGGCACUGGUAGAAUUCACUG Chromosome Location: NA |
| Stem-loop ID | oha-mir-183 |
| Stem-loop Accession # | MI0031380 |
| Stem-loop Sequence | CUGUUCUGUGUAUGGCACUGGUAGAAUUCACUGUGAAAACACUCGAUCAGUGAAUUACAAAGGGCCAUAAACGGAG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0036766
miRBase ID: oha-miR-183-5p Mature miRNA Sequence: UAUGGCACUGGUAGAAUUCACUG Chromosome Location: NA |
| Stem-loop ID | oni-mir-183-1 |
| Stem-loop Accession # | MI0034523 |
| Stem-loop Sequence | UAUGGCACUGGUAGAAUUCACUGUCACAGCACACUAUCAGUGAAUUACCAUAGGGCCAUAAA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0042645
miRBase ID: oni-miR-183 Mature miRNA Sequence: UAUGGCACUGGUAGAAUUCACUG Chromosome Location: NA |
| Stem-loop ID | oni-mir-183-2 |
| Stem-loop Accession # | MI0034522 |
| Stem-loop Sequence | UAUGGCACUGGUAGAAUUCACUGUGAGAGCUCACUAUCAGUGAAUUACCAUAGGGCCAUAAA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0042645
miRBase ID: oni-miR-183 Mature miRNA Sequence: UAUGGCACUGGUAGAAUUCACUG Chromosome Location: NA |
| Stem-loop ID | oni-mir-183-3 |
| Stem-loop Accession # | MI0035058 |
| Stem-loop Sequence | UAUGGCACUGGUAGAAUUCACUGUCACAGCACACUAUCAGUGAAUUACCAUAGGGCCAUA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0042645
miRBase ID: oni-miR-183 Mature miRNA Sequence: UAUGGCACUGGUAGAAUUCACUG Chromosome Location: NA |
| Stem-loop ID | ppa-mir-183 |
| Stem-loop Accession # | MI0003094 |
| Stem-loop Sequence | CCGCAGAGUGUGACUCCUGUUCUGUGUAUGGCACUGGUAGAAUUCACUGUGAACAGUCUCAGUCAGUGAAUUACCGAAGGGCCAUAAACAGAGCAGAGACAGAUCCACGA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0002793
miRBase ID: ppa-miR-183 Mature miRNA Sequence: UAUGGCACUGGUAGAAUUCACUG Chromosome Location: NA |
| Stem-loop ID | ptr-mir-183 |
| Stem-loop Accession # | MI0003086 |
| Stem-loop Sequence | CCGCAGAGUGUGACUCCUGUUCUGUGUAUGGCACUGGUAGAAUUCACUGUGAACAGUCUCAGUCAGUGAAUUACCGAAGGGCCAUAAACAGAGCAGAGACAGAUCCACGA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0002785
miRBase ID: ptr-miR-183 Mature miRNA Sequence: UAUGGCACUGGUAGAAUUCACUG Chromosome Location: NA |
| Stem-loop ID | pma-mir-183 |
| Stem-loop Accession # | MI0017092 |
| Stem-loop Sequence | CUGCCUGCCCUGUCUAUGGCACUGGUAGAAUUCACUGUUCCCGCUGACCUCAGUGAAUUACUAGAGGCCAUAAACAGAGCAGGGAA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0019491
miRBase ID: pma-miR-183-5p Mature miRNA Sequence: UAUGGCACUGGUAGAAUUCACUG Chromosome Location: NA |
| Stem-loop ID | ssc-mir-183 |
| Stem-loop Accession # | MI0002439 |
| Stem-loop Sequence | CUGUGUAUGGCACUGGUAGAAUUCACUGUGAACAGUCUCGGUCAGUGAAUUACCGAAGGGCCAUAAACAG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0002145
miRBase ID: ssc-miR-183 Mature miRNA Sequence: UAUGGCACUGGUAGAAUUCACUG Chromosome Location: NA |
| Stem-loop ID | pny-mir-183-1 |
| Stem-loop Accession # | MI0034795 |
| Stem-loop Sequence | UAUGGCACUGGUAGAAUUCACUGUCACAGCACACUAUCAGUGAAUUACCAUAGGGCCAUAAA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0042855
miRBase ID: pny-miR-183 Mature miRNA Sequence: UAUGGCACUGGUAGAAUUCACUG Chromosome Location: NA |
| Stem-loop ID | pny-mir-183-2 |
| Stem-loop Accession # | MI0034794 |
| Stem-loop Sequence | UAUGGCACUGGUAGAAUUCACUGUGAGAGCUCACUAUCAGUGAAUUACCAUAGGGCCAUAAA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0042855
miRBase ID: pny-miR-183 Mature miRNA Sequence: UAUGGCACUGGUAGAAUUCACUG Chromosome Location: NA |
| Stem-loop ID | pbv-mir-183 |
| Stem-loop Accession # | MI0030260 |
| Stem-loop Sequence | UCCUGUUCUGUGUAUGGCACUGGUAGAAUUCACUGUGAAAACACUCGAUCAGUGAAUUACCAACGGGCCAUAAACGGAGCAGAGAAAGAACC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0038982
miRBase ID: pbv-miR-183-5p Mature miRNA Sequence: UAUGGCACUGGUAGAAUUCACUG Chromosome Location: NA |
| Stem-loop ID | mml-mir-183 |
| Stem-loop Accession # | MI0003084 |
| Stem-loop Sequence | CCGCAGAACGUGACUCCUGUUCUGUGUAUGGCACUGGUAGAAUUCACUGUGAACAGUCUCGGUCAGUGAAUUACCGAAGGGCCAUAAACAGAGCAGAGACAGAUCCACGA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0002783
miRBase ID: mml-miR-183-5p Mature miRNA Sequence: UAUGGCACUGGUAGAAUUCACUG Chromosome Location: NA |
| Stem-loop ID | sla-mir-183 |
| Stem-loop Accession # | MI0003090 |
| Stem-loop Sequence | CCGCAGAACGUGACUCCUGUUCUGUGUAUGGCACUGGUAGAAUUCACUGUGAACAGUCUCGGUCAGUGAAUUACCGAAGGGCCAUAAACAGAGCAGAGACAGAUCCUCGA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0002789
miRBase ID: sla-miR-183 Mature miRNA Sequence: UAUGGCACUGGUAGAAUUCACUG Chromosome Location: NA |
| Stem-loop ID | tni-mir-183 |
| Stem-loop Accession # | MI0003454 |
| Stem-loop Sequence | CUGUGUAUGGCACUGGUAGAAUUCACUGUGAGAGCUCACUAUCAGUGAAUUACCAUAGGGCCAUAAACAG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0003090
miRBase ID: tni-miR-183 Mature miRNA Sequence: UAUGGCACUGGUAGAAUUCACUG Chromosome Location: NA |
| Stem-loop ID | xla-mir-183 |
| Stem-loop Accession # | MI0038274 |
| Stem-loop Sequence | UAUGGCACUGGUAGAAUUCACUGUGAAAACACAAAAUCAGUGAAUUACCAUAGGGCCAUAAA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0046493
miRBase ID: xla-miR-183-5p Mature miRNA Sequence: UAUGGCACUGGUAGAAUUCACUG Chromosome Location: NA |
| Stem-loop ID | xtr-mir-183 |
| Stem-loop Accession # | MI0004852 |
| Stem-loop Sequence | CAUCCCUCCUGUUCUGUGUAUGGCACUGGUAGAAUUCACUGUGAAAACACAAAAUCAGUGAAUUACCAUAGGGCCAUAAACAGAGCAGAGAAAGAAC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0003612
miRBase ID: xtr-miR-183 Mature miRNA Sequence: UAUGGCACUGGUAGAAUUCACUG Chromosome Location: NA |
| Stem-loop ID | dre-mir-183 |
| Stem-loop Accession # | MI0001369 |
| Stem-loop Sequence | GACUCCUGUUCUGUGUAUGGCACUGGUAGAAUUCACUGUGAAAGCACACUAUCAGUGAAUUACCAAAGGGCCAUAAACAGAGCAGAGAAAGAACCACG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0001273
miRBase ID: dre-miR-183-5p Mature miRNA Sequence: UAUGGCACUGGUAGAAUUCACUG Chromosome Location: NA |
More Information
Related Products
miRBase Alias:
Chryseobacterium indologenes: cin-miR-183 (v18)
Danio rerio: dre-miR-183 (v20)
Macaca mulatta: mml-miR-183 (v19)
Monodelphis domestica: mdo-miR-183 (v19)
Phocea massiliensis: pma-miR-183 (v18)
Danio rerio: dre-miR-183 (v20)
Macaca mulatta: mml-miR-183 (v19)
Monodelphis domestica: mdo-miR-183 (v19)
Phocea massiliensis: pma-miR-183 (v18)

Genomic Map