Assay Details
Gene Family ID: | MIPF0000318, mir-488 |
Mature miRNA Sequence: | UUGAAAGGCUAUUUCUUGGUC |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-488-3p | MIMAT0004763 |
| Callithrix jacchus | cja-miR-488 | MIMAT0039524 |
| Dasypus novemcinctus | dno-miR-488-3p | MIMAT0047919 |
| Horse | eca-miR-488 | MIMAT0012965 |
| Pan troglodytes | ptr-miR-488 | MIMAT0008163 |
| Pongo pygmaeus | ppy-miR-488 | MIMAT0015907 |
| View More Rows | ||
| Rabbit | ocu-miR-488-3p | MIMAT0048476 |
| Rhesus monkey | mml-miR-488-3p | MIMAT0006348 |
Stem-loop Details
Human
Callithrix jacchus
Dasypus novemcinctus
Horse
Pan troglodytes
Pongo pygmaeus
Rabbit
Rhesus monkey
More Information
Related Products
miRBase Alias:
Human: hsa-miR-488 (v17)
Macaca mulatta: mml-miR-488 (v19)
Macaca mulatta: mml-miR-488 (v19)

Genomic Map