Assay Details
Amplicon Length: | 87 |
Chromosome Location: | Chr. 1 - 177029765 - 177029765 [+] on 177029765 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-488-5p | MIMAT0002804 |
| Human | hsa-miR-488-3p | MIMAT0004763 |
Stem-loop Details
Human

Genomic Map