Assay Details
Gene Family ID: | MIPF0000185, mir-542 |
Mature miRNA Sequence: | UCGGGGAUCAUCAUGUCACGAGA |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-542-5p | MIMAT0003340 |
| Goat | chi-miR-542-5p | MIMAT0036272 |
| Pongo pygmaeus | ppy-miR-542-5p | MIMAT0015991 |
| Rabbit | ocu-miR-542-5p | MIMAT0048525 |
| Rhesus monkey | mml-miR-542-5p | MIMAT0006406 |
Stem-loop Details
Human
Goat
Pongo pygmaeus
Rabbit
Rhesus monkey

Genomic Map