Assay Details
Amplicon Length: | 99 |
Chromosome Location: | Chr. X - 134541636 - 134541636 [+] on 134541636 Build GRCh38 |
Species: | Human |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-542-5p | MIMAT0003340 |
Human | hsa-miR-542-3p | MIMAT0003389 |
Stem-loop Details
Human
