Assay Details
Gene Family ID: | MIPF0000141, mir-315 |
Mature miRNA Sequence: | UUUUGAUUGUUGCUCAGAAAGCC |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Acyrthosiphon pisum | api-miR-315 | MIMAT0014730 |
| Bactrocera dorsalis | bdo-miR-315 | MIMAT0045390 |
| Capitella teleta | cte-miR-315 | MIMAT0009541 |
| Drosophila virilis | dvi-miR-315a | MIMAT0008944 |
| Lottia gigantea | lgi-miR-315 | MIMAT0009596 |
| Melibe leonina | mle-miR-315-5p | MIMAT0048787 |
| View More Rows | ||
| Triops cancriformis | tcf-miR-315-5p | MIMAT0041404 |
Stem-loop Details
Acyrthosiphon pisum
Bactrocera dorsalis
Capitella teleta
Drosophila virilis
Lottia gigantea
Melibe leonina
Triops cancriformis

Genomic Map