Assay Details
Gene Family ID: | MIPF0000141, mir-315 |
Mature miRNA Sequence: | UUUUGAUUGUUGCUCAGAAAGCC |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Acyrthosiphon pisum | api-miR-315 | MIMAT0014730 |
Bactrocera dorsalis | bdo-miR-315 | MIMAT0045390 |
Capitella teleta | cte-miR-315 | MIMAT0009541 |
Drosophila virilis | dvi-miR-315a | MIMAT0008944 |
Lottia gigantea | lgi-miR-315 | MIMAT0009596 |
Melibe leonina | mle-miR-315-5p | MIMAT0048787 |
View More Rows | ||
Triops cancriformis | tcf-miR-315-5p | MIMAT0041404 |
Stem-loop Details
Acyrthosiphon pisum
Bactrocera dorsalis
Capitella teleta
Drosophila virilis
Lottia gigantea
Melibe leonina
Triops cancriformis
