Assay Details
| Gene Family ID: | MIPF0000141, mir-315 | 
| Mature miRNA Sequence: | UUUUGAUUGUUGCUCUGAGAGUU | 
miRBase Details:
| Species | miRBase ID | miRBase Accession Number | 
|---|---|---|
| Schmidtea mediterranea | sme-miR-315-5p | MIMAT0011282 | 
Stem-loop Details
Schmidtea mediterranea
 Back To Top
 Back To TopMore Information
Related Products
miRBase Alias:
						
							Schmidtea mediterranea: sme-miR-315 (v18)
						
					
				 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top TaqMan™ MicroRNA Assay :
						TaqMan™ MicroRNA Assay :