Assay Details
Gene Family ID: | MIPF0000141, mir-315 |
Mature miRNA Sequence: | UUUUGAUUGUUGCUCUGAGAGUU |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Schmidtea mediterranea | sme-miR-315-5p | MIMAT0011282 |
Stem-loop Details
Schmidtea mediterranea

More Information
Related Products
miRBase Alias:
Schmidtea mediterranea: sme-miR-315 (v18)
