Assay Details
Gene Family ID: | MIPF0001441, mir-3200 |
Mature miRNA Sequence: | CACCUUGCGCUACUCAGGUCUG |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-3200-3p | MIMAT0015085 |
Pan troglodytes | ptr-miR-3200 | MIMAT0024036 |
Rhesus monkey | mml-miR-3200-3p | MIMAT0024323 |
Stem-loop Details
Human
Pan troglodytes
Rhesus monkey
