Assay Details
Gene Family ID: | MIPF0001441, mir-3200 |
Mature miRNA Sequence: | CACCUUGCGCUACUCAGGUCUG |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-3200-3p | MIMAT0015085 |
| Pan troglodytes | ptr-miR-3200 | MIMAT0024036 |
| Rhesus monkey | mml-miR-3200-3p | MIMAT0024323 |
Stem-loop Details
Human
Pan troglodytes
Rhesus monkey

Genomic Map