Assay Details
Gene Family ID: | MIPF0000064, mir-31 |
Mature miRNA Sequence: | CUAUGCCAGCAUCUUGCCU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-31b | MIMAT0017816 |
Stem-loop Details
Rat
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | rno-miR-31b |
|---|---|
| miRBase Accession Number: | MI0015412 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CUAUGCCAGCAUCUUGCCU |
| Species: | Rat |
| Product Type: | mirVana® miRNA mimic |
Gene Family ID: | MIPF0000064, mir-31 |
Mature miRNA Sequence: | CUAUGCCAGCAUCUUGCCU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-31b | MIMAT0017816 |