Assay Details
Amplicon Length: | 62 |
Chromosome Location: | Chr. 5 - 107206868 - 107206868 [+] on 107206868 Build Rnor_6.0 |
Species: | Rat |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-31a-3p | MIMAT0017102 |
| Rat | rno-miR-31a-5p | MIMAT0000810 |
| Rat | rno-miR-31b | MIMAT0017816 |
Stem-loop Details
Rat

Genomic Map