Assay Details
Gene Family ID: | MIPF0000333, mir-652 |
Mature miRNA Sequence: | CAACCCUAGGAGGGGGUGCCAUUC |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-652-5p | MIMAT0017260 |
Stem-loop Details
Mouse
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | mmu-miR-652-5p |
|---|---|
| miRBase Accession Number: | MI0004965 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CAACCCUAGGAGGGGGUGCCAUUC |
| Species: | Mouse |
| Product Type: | mirVana® miRNA mimic |
Gene Family ID: | MIPF0000333, mir-652 |
Mature miRNA Sequence: | CAACCCUAGGAGGGGGUGCCAUUC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-652-5p | MIMAT0017260 |