Assay Details
Amplicon Length: | 69 |
Chromosome Location: | Chr. X - 142738705 - 142738705 [-] on 142738705 Build GRCm38 |
Species: | Mouse |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-652-3p | MIMAT0003711 |
| Mouse | mmu-miR-652-5p | MIMAT0017260 |
Stem-loop Details
Mouse

Genomic Map