Assay Details
Gene Family ID: | MIPF0000316, mir-467 |
Mature miRNA Sequence: | AUACAUACACACACACAUACAC |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-466b-3p | MIMAT0017285 |
Stem-loop Details
Rat
More Information
Related Products
miRBase Alias:
Rat: rno-miR-466b-1-3p (v20)
Rat: rno-miR-466b-1* (v18)
Rat: rno-miR-466b-1* (v18)

Genomic Map