Assay Details
Amplicon Length: | 90 |
Chromosome Location: | Chr. 17 - 72069313 - 72069313 [-] on 72069313 Build Rnor_6.0 |
Species: | Rat |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-466b-5p | MIMAT0005278 |
| Rat | rno-miR-466b-3p | MIMAT0017285 |
Stem-loop Details
Rat

Genomic Map