Assay Details
Gene Family ID: | MIPF0000018, mir-154 |
Mature miRNA Sequence: | AGAGGUUAUCCGUGUUAUGUUC |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-655-5p | MIMAT0026626 |
| Sheep | oar-miR-655-5p | MIMAT0019300 |
Stem-loop Details
Human
Sheep

Genomic Map