Assay Details
| Gene Family ID: | MIPF0000254, mir-316 | 
| Mature miRNA Sequence: | UGUCUUUUUCCGCUUUGCUGC | 
miRBase Details:
| Species | miRBase ID | miRBase Accession Number | 
|---|---|---|
| Heliconius melpomene | hme-miR-316 | MIMAT0024978 | 
| Tribolium castaneum | tca-miR-316-5p | MIMAT0018716 | 
Stem-loop Details
Heliconius melpomene
Tribolium castaneum
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top TaqMan™ MicroRNA Assay :
						TaqMan™ MicroRNA Assay :