Assay Details
Gene Family ID: | MIPF0000254, mir-316 |
Mature miRNA Sequence: | UGUCUUUUUCCGCUUUGCUGC |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Heliconius melpomene | hme-miR-316 | MIMAT0024978 |
| Tribolium castaneum | tca-miR-316-5p | MIMAT0018716 |
Stem-loop Details
Heliconius melpomene
Tribolium castaneum

Genomic Map