Assay Details
Gene Family ID: | MIPF0000372, mir-392 |
Mature miRNA Sequence: | AGCAUUCGUGGUUGAGGAUAU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Caenorhabditis elegans | cel-miR-392-5p | MIMAT0020344 |
Stem-loop Details
Caenorhabditis elegans
More Information
Related Products
miRBase Alias:
Caenorhabditis elegans: cel-miR-392* (v17)

Genomic Map