Assay Details
Gene Family ID: | MIPF0000372, mir-392 |
Mature miRNA Sequence: | AGCAUUCGUGGUUGAGGAUAU |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Caenorhabditis elegans | cel-miR-392-5p | MIMAT0020344 |
Stem-loop Details
Caenorhabditis elegans

More Information
Related Products
miRBase Alias:
Caenorhabditis elegans: cel-miR-392* (v17)
